Product Details

SNP ID
rs201006442
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:42851253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAACCTTCAATAATACCTCCAAC[C/G]TGAGAACACACCAGCGGATCCACAC
Phenotype
MIM: 609017
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERMAP PubMed Links

Gene Details

Gene
ERMAP
Gene Name
erythroblast membrane associated protein (Scianna blood group)
There are no transcripts associated with this gene.

Gene
LOC339539
Gene Name
uncharacterized LOC339539
There are no transcripts associated with this gene.

Gene
ZNF691
Gene Name
zinc finger protein 691
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242739.1 4372 Silent Mutation CTG,GTG L130V NP_001229668.1
NM_015911.3 4372 Silent Mutation CTG,GTG L99V NP_056995.1
XM_006710671.3 4372 Missense Mutation CTG,GTG L126V XP_006710734.1
XM_017001400.1 4372 Missense Mutation CTG,GTG L130V XP_016856889.1
XM_017001401.1 4372 Missense Mutation CTG,GTG L130V XP_016856890.1
XM_017001402.1 4372 Missense Mutation CTG,GTG L121V XP_016856891.1
XM_017001403.1 4372 Missense Mutation CTG,GTG L121V XP_016856892.1

View Full Product Details