Product Details

SNP ID
rs201169467
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:162381962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCAGGCCAGCCGCTCCTTCCCC[A/C]ACTTGTCCAAGCCCGTGGCCCCCGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf111 PubMed Links

Gene Details

Gene
C1orf111
Gene Name
chromosome 1 open reading frame 111
There are no transcripts associated with this gene.

Gene
C1orf226
Gene Name
chromosome 1 open reading frame 226
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085375.1 233 Missense Mutation AAC,CAC N21H NP_001078844.1
NM_001135240.1 233 Missense Mutation AAC,CAC N64H NP_001128712.1

View Full Product Details