Product Details

SNP ID
rs201516481
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:155321873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTCCCGCCGTCCTGAGCTACAG[C/G]AGGGGCCTTTGAGCACACCCCCTCC
Phenotype
MIM: 134629
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FDPS PubMed Links

Gene Details

Gene
FDPS
Gene Name
farnesyl diphosphate synthase
There are no transcripts associated with this gene.

Gene
LOC105371451
Gene Name
uncharacterized LOC105371451
There are no transcripts associated with this gene.

Gene
RUSC1
Gene Name
RUN and SH3 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105203.1 329 Missense Mutation CAG,GAG Q34E NP_001098673.1
NM_001105204.1 329 Missense Mutation CAG,GAG Q34E NP_001098674.1
NM_001105205.1 329 Intron NP_001098675.1
NM_001278227.1 329 Intron NP_001265156.1
NM_001278228.1 329 Intron NP_001265157.1
NM_001278229.1 329 Intron NP_001265158.1
NM_001278230.1 329 Intron NP_001265159.1
NM_014328.4 329 Intron NP_055143.2
XM_006711254.1 329 Missense Mutation CAG,GAG Q34E XP_006711317.1
XM_006711256.1 329 Missense Mutation CAG,GAG Q34E XP_006711319.1
XM_006711257.1 329 Intron XP_006711320.1
XM_017000891.1 329 Missense Mutation CAG,GAG Q34E XP_016856380.1
XM_017000892.1 329 Intron XP_016856381.1
Gene
RUSC1-AS1
Gene Name
RUSC1 antisense RNA 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039517.1 329 Intron NP_001034606.1

View Full Product Details