Product Details

SNP ID
rs200095317
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:88857290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAGATGGGGCTCCTCCCAGGAA[C/T]CAAGTCCAGTTCCACAGCTTTACGG
Phenotype
MIM: 189963
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTF2B PubMed Links

Gene Details

Gene
GTF2B
Gene Name
general transcription factor IIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001514.5 1054 Missense Mutation ATT,GTT I245V NP_001505.1
XM_011541299.1 1054 Missense Mutation ATT,GTT I214V XP_011539601.1

View Full Product Details