Product Details

SNP ID
rs199545183
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:219968889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCTCCAGGCTGCAGTTCACAGA[A/G]TGGTTTGAAGGGGATGCAAAGGCTT
Phenotype
MIM: 138295 MIM: 611146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPRS PubMed Links

Gene Details

Gene
EPRS
Gene Name
glutamyl-prolyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004446.2 2982 Missense Mutation CTC,TTC L1486F NP_004437.2
XM_017000614.1 2982 Missense Mutation CTC,TTC L960F XP_016856103.1
Gene
SLC30A10
Gene Name
solute carrier family 30 member 10
There are no transcripts associated with this gene.

View Full Product Details