Product Details

SNP ID
rs199891634
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:151364678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCCCTCCTTTTACTGTGTCTAC[A/G]TCAACCTGCAGCATCACAGAGCCTT
Phenotype
MIM: 604188
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SELENBP1 PubMed Links

Gene Details

Gene
SELENBP1
Gene Name
selenium binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258288.1 1586 Silent Mutation GAC,GAT D366D NP_001245217.1
NM_001258289.1 1586 Silent Mutation GAC,GAT D470D NP_001245218.1
NM_003944.3 1586 Silent Mutation GAC,GAT D428D NP_003935.2
XM_017002757.1 1586 Intron XP_016858246.1

View Full Product Details