Product Details

SNP ID
rs199922959
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:206930335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACCGTCTAGGCTTCCTGGGGGC[C/T]GTCCTGGGCCTCGGCGGCCACGGTG
Phenotype
MIM: 606015 MIM: 173880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCMR PubMed Links

Gene Details

Gene
FCMR
Gene Name
Fc fragment of IgM receptor
There are no transcripts associated with this gene.

Gene
PIGR
Gene Name
polymeric immunoglobulin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002644.3 2462 Missense Mutation AGC,GGC S760G NP_002635.2
XM_011509629.1 2462 Missense Mutation AGC,GGC S760G XP_011507931.1

View Full Product Details