Product Details

SNP ID
rs199963724
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:212624929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATACGGCCCAGAGTGGCTCCAGC[A/G]TGAGCATGTTCAACACCACCATGGG
Phenotype
MIM: 603148
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATF3 PubMed Links

Gene Details

Gene
ATF3
Gene Name
activating transcription factor 3
There are no transcripts associated with this gene.

Gene
FAM71A
Gene Name
family with sequence similarity 71 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153606.3 483 Missense Mutation ATG,GTG M18V NP_705834.2

View Full Product Details