Product Details

SNP ID
rs199724441
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:53506658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGGAGGGGATGTGGCCCAGGCA[A/G]TGGTCAAAGGCTCCATTGGGGAAGA
Phenotype
MIM: 610378
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLIS1 PubMed Links

Gene Details

Gene
GLIS1
Gene Name
GLIS family zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147193.2 2111 Silent Mutation CAC,CAT H608H NP_671726.2
XM_017000408.1 2111 Silent Mutation CAC,CAT H783H XP_016855897.1
XM_017000409.1 2111 Silent Mutation CAC,CAT H779H XP_016855898.1
XM_017000410.1 2111 Silent Mutation CAC,CAT H783H XP_016855899.1
XM_017000411.1 2111 UTR 3 XP_016855900.1
XM_017000412.1 2111 Intron XP_016855901.1

View Full Product Details