Product Details

SNP ID
rs199592567
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:34988898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTCTTTGAAGGTGGAGAATC[A/G]AGTTCTGCATCATTTTTTTCATTAG
Phenotype
MIM: 613567
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM35B PubMed Links

Gene Details

Gene
TMEM35B
Gene Name
transmembrane protein 35B
There are no transcripts associated with this gene.

Gene
ZMYM6
Gene Name
zinc finger MYM-type containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007167.3 2412 Silent Mutation CTC,CTT L728L NP_009098.3

View Full Product Details