Product Details
- SNP ID
-
rs200094994
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:31619302 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGAGTTTCTCCGCTATCTGTGGC[A/G]CGATTATCTGTACCCAAAACAGTAT
- Phenotype
-
MIM: 602392
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HCRTR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1056526] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HCRTR1
- Gene Name
- hypocretin receptor 1
View Full Product Details