Product Details

SNP ID
rs199517535
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:11922649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCTTCCTACCTTACTCCTGAAG[A/T]GGGGCTTGGCTCCAGGCCCACAGTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA2013 PubMed Links

Gene Details

Gene
KIAA2013
Gene Name
KIAA2013
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138346.2 2065 Missense Mutation CAC,CTC H625L NP_612355.1
XM_005263531.4 2065 Missense Mutation CAC,CTC H625L XP_005263588.1

View Full Product Details