Product Details
- SNP ID
-
rs200261270
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:152107697 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCCGCCCGGTACTGCCGGTCTCG[C/G]TCCTGCCGCAGCCTCTGCTCTTGTT
- Phenotype
-
MIM: 190370
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TCHH
PubMed Links
Gene Details
- Gene
- TCHH
- Gene Name
- trichohyalin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007113.3 |
5615 |
Missense Mutation |
GAC,GAG |
D1840E |
NP_009044.2 |
View Full Product Details