Product Details

SNP ID
rs200340103
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:207090389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAAGGACAGATTCTGGGGACTT[A/T]CGTTTGTATCAAGGGCTACCACCTG
Phenotype
MIM: 120831 MIM: 171835
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C4BPB PubMed Links

Gene Details

Gene
C4BPB
Gene Name
complement component 4 binding protein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000716.3 430 Missense Mutation TAC,TTC Y47F NP_000707.1
NM_001017364.1 430 Missense Mutation TAC,TTC Y46F NP_001017364.1
NM_001017365.2 430 Missense Mutation TAC,TTC Y47F NP_001017365.1
NM_001017366.2 430 Missense Mutation TAC,TTC Y46F NP_001017366.1
NM_001017367.1 430 Missense Mutation TAC,TTC Y47F NP_001017367.1
XM_005273254.4 430 Missense Mutation TAC,TTC Y47F XP_005273311.1
XM_005273255.2 430 Missense Mutation TAC,TTC Y46F XP_005273312.1
Gene
PFKFB2
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2
There are no transcripts associated with this gene.

View Full Product Details