Product Details

SNP ID
rs200685743
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:156729151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCTCTCATGAGGGGAGGAAGC[A/G]GCTAGCTGTTAACCTCACCCGTGTC
Phenotype
MIM: 611930 MIM: 611836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ISG20L2 PubMed Links

Gene Details

Gene
ISG20L2
Gene Name
interferon stimulated exonuclease gene 20 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303095.1 254 Intron NP_001290024.1
XM_005245516.4 254 Intron XP_005245573.1
XM_005245518.4 254 Intron XP_005245575.1
XM_006711560.3 254 Intron XP_006711623.1
XM_006711561.3 254 Intron XP_006711624.1
Gene
MRPL24
Gene Name
mitochondrial ribosomal protein L24
There are no transcripts associated with this gene.

Gene
RRNAD1
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142560.1 254 Missense Mutation CAG,CGG Q16R NP_001136032.1
NM_015997.3 254 Missense Mutation CAG,CGG Q16R NP_057081.3
XM_005245234.4 254 Missense Mutation CAG,CGG Q16R XP_005245291.1
XM_005245235.4 254 Missense Mutation CAG,CGG Q16R XP_005245292.1
XM_005245236.4 254 Missense Mutation CAG,CGG Q16R XP_005245293.1
XM_005245237.4 254 Missense Mutation CAG,CGG Q16R XP_005245294.1
XM_011509600.1 254 Intron XP_011507902.1
XM_017001404.1 254 UTR 5 XP_016856893.1
XM_017001405.1 254 UTR 5 XP_016856894.1
XM_017001406.1 254 UTR 5 XP_016856895.1

View Full Product Details