Product Details
- SNP ID
-
rs200368420
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:20807406 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGAGCCACGCCCTTCCCATTCTG[C/T]TCTGCAGGGTCCTTGCTGCTCTCCC
- Phenotype
-
MIM: 603929
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EIF4G3
PubMed Links
Gene Details
- Gene
- EIF4G3
- Gene Name
- eukaryotic translation initiation factor 4 gamma 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001198801.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1593E |
NP_001185730.1 |
NM_001198802.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1563E |
NP_001185731.1 |
NM_001198803.2 |
5779 |
Intron |
|
|
NP_001185732.1 |
NM_003760.4 |
5779 |
Silent Mutation |
GAA,GAG |
E1557E |
NP_003751.2 |
XM_011542356.2 |
5779 |
Silent Mutation |
GAA,GAG |
E1649E |
XP_011540658.1 |
XM_011542361.2 |
5779 |
Silent Mutation |
GAA,GAG |
E1612E |
XP_011540663.1 |
XM_011542370.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1650E |
XP_011540672.1 |
XM_011542371.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1605E |
XP_011540673.1 |
XM_011542374.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1594E |
XP_011540676.1 |
XM_011542375.2 |
5779 |
Silent Mutation |
GAA,GAG |
E1594E |
XP_011540677.1 |
XM_011542381.2 |
5779 |
Silent Mutation |
GAA,GAG |
E1109E |
XP_011540683.1 |
XM_017002672.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1650E |
XP_016858161.1 |
XM_017002673.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1643E |
XP_016858162.1 |
XM_017002674.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1642E |
XP_016858163.1 |
XM_017002675.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1639E |
XP_016858164.1 |
XM_017002676.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1632E |
XP_016858165.1 |
XM_017002677.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1632E |
XP_016858166.1 |
XM_017002678.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1613E |
XP_016858167.1 |
XM_017002679.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1612E |
XP_016858168.1 |
XM_017002680.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1612E |
XP_016858169.1 |
XM_017002681.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1611E |
XP_016858170.1 |
XM_017002682.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1605E |
XP_016858171.1 |
XM_017002683.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1605E |
XP_016858172.1 |
XM_017002684.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1605E |
XP_016858173.1 |
XM_017002685.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1602E |
XP_016858174.1 |
XM_017002686.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1601E |
XP_016858175.1 |
XM_017002687.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1601E |
XP_016858176.1 |
XM_017002688.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1601E |
XP_016858177.1 |
XM_017002689.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1601E |
XP_016858178.1 |
XM_017002690.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1600E |
XP_016858179.1 |
XM_017002691.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1595E |
XP_016858180.1 |
XM_017002692.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1594E |
XP_016858181.1 |
XM_017002693.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1594E |
XP_016858182.1 |
XM_017002694.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1575E |
XP_016858183.1 |
XM_017002695.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1574E |
XP_016858184.1 |
XM_017002696.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1568E |
XP_016858185.1 |
XM_017002697.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1567E |
XP_016858186.1 |
XM_017002698.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1564E |
XP_016858187.1 |
XM_017002699.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1564E |
XP_016858188.1 |
XM_017002700.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1557E |
XP_016858189.1 |
XM_017002701.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1556E |
XP_016858190.1 |
XM_017002702.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1534E |
XP_016858191.1 |
XM_017002703.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1509E |
XP_016858192.1 |
XM_017002704.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1508E |
XP_016858193.1 |
XM_017002705.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1370E |
XP_016858194.1 |
XM_017002706.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1333E |
XP_016858195.1 |
XM_017002707.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1314E |
XP_016858196.1 |
XM_017002708.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1277E |
XP_016858197.1 |
XM_017002709.1 |
5779 |
Silent Mutation |
GAA,GAG |
E1149E |
XP_016858198.1 |
XM_017002710.1 |
5779 |
Intron |
|
|
XP_016858199.1 |
XM_017002711.1 |
5779 |
Intron |
|
|
XP_016858200.1 |
XM_017002712.1 |
5779 |
Intron |
|
|
XP_016858201.1 |
View Full Product Details