Product Details

SNP ID
rs200409312
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:229627634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAATTTTAGATAAAGCCTAGCC[A/G]TGGCTGCTGTTTATTCTGGCATTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAF5L PubMed Links

Gene Details

Gene
TAF5L
Gene Name
TATA-box binding protein associated factor 5 like
There are no transcripts associated with this gene.

Gene
URB2
Gene Name
URB2 ribosome biogenesis 2 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314021.1 155 Missense Mutation ATG,GTG M1V NP_001300950.1
NM_014777.3 155 Missense Mutation ATG,GTG M1V NP_055592.2
XM_005273360.3 155 Missense Mutation ATG,GTG M1V XP_005273417.1

View Full Product Details