Product Details
- SNP ID
-
rs200729611
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:246544549 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTGGTCTATTACAGTGGCGCTGC[A/G]CCTCCCAAAACAGTGCTTTAACAAG
- Phenotype
-
MIM: 607055
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TFB2M
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3129568] are located under a probe and SNP(s) [rs3124119] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TFB2M
- Gene Name
- transcription factor B2, mitochondrial
View Full Product Details