Product Details

SNP ID
rs200390008
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:247451198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCTGGAAGGGGGCTGCAGATAC[A/C]TGTAAATCGCAGGTAGGTAGAAGAG
Phenotype
MIM: 606416
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NLRP3 PubMed Links

Gene Details

Gene
NLRP3
Gene Name
NLR family pyrin domain containing 3
There are no transcripts associated with this gene.

Gene
OR2B11
Gene Name
olfactory receptor family 2 subfamily B member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004492.1 785 Missense Mutation AGG,ATG R262M NP_001004492.1

View Full Product Details