Product Details

SNP ID
rs200323920
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:84813943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTGAGGACTGTGGAGGGGATG[A/C]GAGAGGGACGCCTCTCTGGGTTCTC
Phenotype
MIM: 605106
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LPAR3 PubMed Links

Gene Details

Gene
LPAR3
Gene Name
lysophosphatidic acid receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012152.2 1181 Missense Mutation NP_036284.1

View Full Product Details