Product Details

SNP ID
rs200190849
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:203483445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCCTGATTTCCCATCTGCCCT[C/G]TACTGTGATAGCCGCAACCTGCGAA
Phenotype
MIM: 601914
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRELP PubMed Links

Gene Details

Gene
PRELP
Gene Name
proline and arginine rich end leucine rich repeat protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002725.3 461 Silent Mutation CTC,CTG L87L NP_002716.1
NM_201348.1 461 Silent Mutation CTC,CTG L87L NP_958505.1

View Full Product Details