Product Details

SNP ID
rs200165834
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:151176714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGGGGCTCAGGAGGTGGCGGG[C/T]CGGGGCCCAGGGCTGAGGCAGTGGG
Phenotype
MIM: 608095 MIM: 605834 MIM: 600607
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCNM1 PubMed Links

Gene Details

Gene
SCNM1
Gene Name
sodium channel modifier 1
There are no transcripts associated with this gene.

Gene
TMOD4
Gene Name
tropomodulin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013353.2 1117 Intron NP_037485.2
XM_011509449.1 1117 Intron XP_011507751.1
XM_017001089.1 1117 Intron XP_016856578.1
XM_017001090.1 1117 Intron XP_016856579.1
Gene
TNFAIP8L2-SCNM1
Gene Name
TNFAIP8L2-SCNM1 readthrough
There are no transcripts associated with this gene.

Gene
VPS72
Gene Name
vacuolar protein sorting 72 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271087.1 1117 Missense Mutation GAC,GGC D353G NP_001258016.1
NM_001271088.1 1117 UTR 3 NP_001258017.1
NM_005997.2 1117 Missense Mutation GAC,GGC D342G NP_005988.1
XM_017002205.1 1117 Missense Mutation GAC,GGC D221G XP_016857694.1

View Full Product Details