Product Details

SNP ID
rs200206735
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:151364583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTACAATCGCCCCCAGGGTAG[C/T]GGAGCTCATGGGCAAGGGCTGGGCC
Phenotype
MIM: 604188
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SELENBP1 PubMed Links

Gene Details

Gene
SELENBP1
Gene Name
selenium binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258288.1 1681 Missense Mutation CAC,CGC H398R NP_001245217.1
NM_001258289.1 1681 Missense Mutation CAC,CGC H502R NP_001245218.1
NM_003944.3 1681 Missense Mutation CAC,CGC H460R NP_003935.2
XM_017002757.1 1681 Intron XP_016858246.1

View Full Product Details