Product Details
- SNP ID
-
rs202022682
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:3816284 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAGCGGCGCTGTCCCTCACCTGG[C/T]TGCAGTGCCGGGGCCTTCTGCAGAA
- Phenotype
-
MIM: 616690
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CEP104
PubMed Links
Gene Details
- Gene
- CEP104
- Gene Name
- centrosomal protein 104
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014704.3 |
3119 |
Silent Mutation |
CAA,CAG |
Q886Q |
NP_055519.1 |
XM_005244815.4 |
3119 |
Silent Mutation |
CAA,CAG |
Q922Q |
XP_005244872.1 |
XM_011542473.2 |
3119 |
Silent Mutation |
CAA,CAG |
Q928Q |
XP_011540775.1 |
XM_011542474.2 |
3119 |
Silent Mutation |
CAA,CAG |
Q892Q |
XP_011540776.1 |
XM_011542475.2 |
3119 |
Silent Mutation |
CAA,CAG |
Q870Q |
XP_011540777.1 |
XM_011542476.2 |
3119 |
Silent Mutation |
CAA,CAG |
Q862Q |
XP_011540778.1 |
XM_011542477.2 |
3119 |
Silent Mutation |
CAA,CAG |
Q804Q |
XP_011540779.1 |
XM_011542478.1 |
3119 |
Intron |
|
|
XP_011540780.1 |
XM_017002917.1 |
3119 |
Silent Mutation |
CAA,CAG |
Q864Q |
XP_016858406.1 |
XM_017002918.1 |
3119 |
Silent Mutation |
CAA,CAG |
Q828Q |
XP_016858407.1 |
XM_017002919.1 |
3119 |
Silent Mutation |
CAA,CAG |
Q820Q |
XP_016858408.1 |
View Full Product Details