Product Details

SNP ID
rs201879768
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:229325350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGAGTGAGCTCGCTGCCTTTGA[C/G]CAACCAGTTTCCTTTTTTCCACCTG
Phenotype
MIM: 616762
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCSAP PubMed Links

Gene Details

Gene
CCSAP
Gene Name
centriole, cilia and spindle associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145257.4 776 Missense Mutation GCT,GGT A233G NP_660300.3

View Full Product Details