Product Details

SNP ID
rs201959823
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:156312066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGAGAATCTCTTTGCTAGCCCCC[C/T]GGAGGAGAATGGTGCAGGCCTTGGG
Phenotype
MIM: 600114
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCT3 PubMed Links

Gene Details

Gene
CCT3
Gene Name
chaperonin containing TCP1 subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008800.2 1247 Missense Mutation NP_001008800.1
NM_005998.4 1247 Missense Mutation NP_005989.3

View Full Product Details