Product Details

SNP ID
rs201893746
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:42767533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCCTGGTGGAGCTCACCCCGA[A/C]CCCCGGCGGCCTGGCCCTGGTGAGC
Phenotype
MIM: 610339
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf50 PubMed Links

Gene Details

Gene
C1orf50
Gene Name
chromosome 1 open reading frame 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024097.3 171 Missense Mutation AAC,ACC N35T NP_077002.2
Gene
LOC105378683
Gene Name
uncharacterized LOC105378683
There are no transcripts associated with this gene.

Gene
P3H1
Gene Name
prolyl 3-hydroxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146289.1 171 Intron NP_001139761.1
NM_001243246.1 171 Intron NP_001230175.1
NM_022356.3 171 Intron NP_071751.3
XM_005271110.2 171 Intron XP_005271167.1
XM_011541948.1 171 Intron XP_011540250.1
XM_017002051.1 171 Intron XP_016857540.1
XM_017002052.1 171 Intron XP_016857541.1

View Full Product Details