Product Details

SNP ID
rs201881922
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:31619366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCAGCCTATGTGGCTGTGTTCGT[C/T]GTGGCCCTGGTGGGCAACACGCTGG
Phenotype
MIM: 602392
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCRTR1 PubMed Links

Gene Details

Gene
HCRTR1
Gene Name
hypocretin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001525.2 1050 Silent Mutation GTC,GTT V58V NP_001516.2
XM_017001105.1 1050 Silent Mutation GTC,GTT V58V XP_016856594.1
XM_017001106.1 1050 Silent Mutation GTC,GTT V58V XP_016856595.1
XM_017001107.1 1050 Silent Mutation GTC,GTT V58V XP_016856596.1

View Full Product Details