Product Details

SNP ID
rs202003978
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:26041758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCAAGGTTGTCCCCAGGACCAG[G/T]ATGGAGAAGACGAAGGTGCAGATGG
Phenotype
MIM: 601738 MIM: 609617 MIM: 606131
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EXTL1 PubMed Links

Gene Details

Gene
EXTL1
Gene Name
exostosin like glycosyltransferase 1
There are no transcripts associated with this gene.

Gene
SLC30A2
Gene Name
solute carrier family 30 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004434.2 1022 Silent Mutation NP_001004434.1
NM_032513.4 1022 Silent Mutation NP_115902.1
Gene
TRIM63
Gene Name
tripartite motif containing 63
There are no transcripts associated with this gene.

View Full Product Details