Product Details

SNP ID
rs202069770
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:228101222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTCCTCTTACAGCAGGGGGAG[C/T]TGGAGTCGGAGTCATGGTGGTGGTG
Phenotype
MIM: 103180 MIM: 611859
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARF1 PubMed Links

Gene Details

Gene
ARF1
Gene Name
ADP ribosylation factor 1
There are no transcripts associated with this gene.

Gene
C1orf35
Gene Name
chromosome 1 open reading frame 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024319.3 936 Missense Mutation ACC,ATC T234I NP_077295.1
Gene
MIR3620
Gene Name
microRNA 3620
There are no transcripts associated with this gene.

Gene
MRPL55
Gene Name
mitochondrial ribosomal protein L55
There are no transcripts associated with this gene.

View Full Product Details