Product Details

SNP ID
rs199906203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46009902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTATGGTTACACTCGGGTGGCA[C/G]AGATGCGTGGAGAGTCGAAATCTCT
Phenotype
MIM: 120361
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MMP9 PubMed Links

Gene Details

Gene
MMP9
Gene Name
matrix metallopeptidase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004994.2 194 Missense Mutation CAG,GAG Q59E NP_004985.2

View Full Product Details