Product Details

SNP ID
rs200013287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:22582768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCTTTGCGTGCGTGTAGCTGCG[A/C]CTGTAGGTCTTGGGGTCGCGGGCGC
Phenotype
MIM: 600288
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FOXA2 PubMed Links

Gene Details

Gene
FOXA2
Gene Name
forkhead box A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021784.4 646 Missense Mutation AGG,AGT R158S NP_068556.2
NM_153675.2 646 Missense Mutation AGG,AGT R152S NP_710141.1
Gene
LINC00261
Gene Name
long intergenic non-protein coding RNA 261
There are no transcripts associated with this gene.

Gene
LINC01384
Gene Name
long intergenic non-protein coding RNA 1384
There are no transcripts associated with this gene.

View Full Product Details