Product Details

SNP ID
rs199667415
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37301354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGTACGGACTCTTCCTGGGAGCC[A/G]TCTTCCAGCTCATCTGTGTGCTGGC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MANBAL PubMed Links

Gene Details

Gene
MANBAL
Gene Name
mannosidase beta like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003897.1 1361 Missense Mutation ATC,GTC I31V NP_001003897.1
NM_022077.3 1361 Missense Mutation ATC,GTC I31V NP_071360.1
XM_017028006.1 1361 Missense Mutation ATC,GTC I31V XP_016883495.1
XM_017028007.1 1361 Missense Mutation ATC,GTC I31V XP_016883496.1

View Full Product Details