Product Details

SNP ID
rs201828251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:52973324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGAAAACAGCAGGCACCCAAG[A/C]GGGCGGCAGGTAAGAGAAACGGCTC
Phenotype
MIM: 614118
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TSHZ2 PubMed Links

Gene Details

Gene
TSHZ2
Gene Name
teashirt zinc finger homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193421.1 1016 Intron NP_001180350.1
NM_173485.5 1016 Silent Mutation AGG,CGG R11R NP_775756.3
XM_017027640.1 1016 Silent Mutation AGG,CGG R11R XP_016883129.1
XM_017027641.1 1016 Silent Mutation AGG,CGG R11R XP_016883130.1

View Full Product Details