Product Details

SNP ID
rs201926200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63559315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCACGAGGGTCTGCTGAGCCTC[A/G]CAGAAGTCCAGGAGGCTACGCCAGA
Phenotype
MIM: 611265
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FNDC11 PubMed Links

Gene Details

Gene
FNDC11
Gene Name
fibronectin type III domain containing 11
There are no transcripts associated with this gene.

Gene
HELZ2
Gene Name
helicase with zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037335.2 8773 Silent Mutation TGC,TGT C2627C NP_001032412.2
NM_033405.3 8773 Silent Mutation TGC,TGT C2058C NP_208384.3

View Full Product Details