Product Details
- SNP ID
-
rs201877331
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23750671 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGCTCTTAGTACCCGCAGCGGAC[A/G]TCTGTAGTAGTCATCTTTGGTGGCC
- Phenotype
-
MIM: 123855
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CST1
PubMed Links
Gene Details
- Gene
- CST1
- Gene Name
- cystatin SN
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001898.2 |
267 |
Missense Mutation |
AGT,GGT |
S66G |
NP_001889.2 |
View Full Product Details