Product Details
- SNP ID
-
rs201692282
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:56249250 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGTGGCCTCCATCTGCAACCTCC[C/T]GGCCATCGCCGTCGACAGGTACGTC
- Phenotype
-
MIM: 155540
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MC3R
PubMed Links
Gene Details
- Gene
- MC3R
- Gene Name
- melanocortin 3 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019888.3 |
519 |
Missense Mutation |
CCG,CTG |
P136L |
NP_063941.3 |
View Full Product Details