Product Details

SNP ID
rs200536017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56395659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGCATCGCCAATGGCCTCATCA[A/G]TGAAATCAAGCCTCAGTCTGTGTGT
Phenotype
MIM: 603072 MIM: 600369
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AURKA PubMed Links

Gene Details

Gene
AURKA
Gene Name
aurora kinase A
There are no transcripts associated with this gene.

Gene
CSTF1
Gene Name
cleavage stimulation factor subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033521.1 307 Missense Mutation AAT,AGT N36S NP_001028693.1
NM_001033522.1 307 Missense Mutation AAT,AGT N36S NP_001028694.1
NM_001324.2 307 Missense Mutation AAT,AGT N36S NP_001315.1
XM_011528600.1 307 Missense Mutation AAT,AGT N36S XP_011526902.1

View Full Product Details