Product Details

SNP ID
rs200297144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23750640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGGGTGGAGGGAGCACCTACC[A/T]GTTGCCTGGCTCTTAGTACCCGCAG
Phenotype
MIM: 123855
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CST1 PubMed Links

Gene Details

Gene
CST1
Gene Name
cystatin SN
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001898.2 298 Missense Mutation CAG,CTG Q76L NP_001889.2

View Full Product Details