Product Details

SNP ID
rs201109202
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3854629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCAGAATCTGAGCAGCAATGC[C/T]GTTTGCTGAAGACAAGACCTATAAG
Phenotype
MIM: 609676
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MAVS PubMed Links

Gene Details

Gene
MAVS
Gene Name
mitochondrial antiviral signaling protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206491.1 177 UTR 5 NP_001193420.1
NM_020746.4 177 Missense Mutation CCG,CTG P2L NP_065797.2

View Full Product Details