Product Details

SNP ID
rs200707079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35003473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCTGCTGCCAGAAGCGCAGCAG[A/G]TTGTGGAAGTTGTTGAGCAGGAAGC
Phenotype
MIM: 609928 MIM: 608430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYH7B PubMed Links

Gene Details

Gene
MYH7B
Gene Name
myosin heavy chain 7B
There are no transcripts associated with this gene.

Gene
TRPC4AP
Gene Name
transient receptor potential cation channel subfamily C member 4 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015638.2 1705 Silent Mutation NP_056453.1
NM_199368.1 1705 Silent Mutation NP_955400.1
XM_011528772.2 1705 Intron XP_011527074.1
XM_011528774.1 1705 Silent Mutation XP_011527076.1
XM_017027799.1 1705 Intron XP_016883288.1
XM_017027800.1 1705 Silent Mutation XP_016883289.1
XM_017027801.1 1705 Silent Mutation XP_016883290.1

View Full Product Details