Product Details
- SNP ID
-
rs200307238
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.20:23750782 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGTTATAGATGCCACCCGGGATTA[C/T]CCTATCCTCCTCCTTGGGGCTCCAG
- Phenotype
-
MIM: 123855
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CST1
PubMed Links
Gene Details
- Gene
- CST1
- Gene Name
- cystatin SN
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001898.2 |
156 |
Missense Mutation |
ATA,GTA |
I29V |
NP_001889.2 |
View Full Product Details