Product Details

SNP ID
rs201422446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23633922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGCCCGGTACAGACCCCTAGGC[A/G]TCCTGACAGGTGGATTTCGACAAGG
Phenotype
MIM: 604312
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CST3 PubMed Links

Gene Details

Gene
CST3
Gene Name
cystatin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000099.3 621 Silent Mutation GAC,GAT D145D NP_000090.1
NM_001288614.1 621 Silent Mutation GAC,GAT D145D NP_001275543.1

View Full Product Details