Product Details

SNP ID
rs201397350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32603208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTTTTTTCATTCTTCCCCACGT[A/T]GTCTGAAAGCTTCTTCGTTCTTCTC
Phenotype
MIM: 615494
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C21orf59 PubMed Links

Gene Details

Gene
C21orf59
Gene Name
chromosome 21 open reading frame 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021254.2 984 Missense Mutation AAC,TAC N207Y NP_067077.1

View Full Product Details