Product Details

SNP ID
rs199473363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34370537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGACGTCTTCCGAAGGATTTTTA[A/T]TACTTATATGGACAATTGGCGCCAG
Phenotype
MIM: 603796
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNE2 PubMed Links

Gene Details

Gene
KCNE2
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172201.1 199 Missense Mutation AAT,ATT N20I NP_751951.1
Gene
SMIM11A
Gene Name
small integral membrane protein 11A
There are no transcripts associated with this gene.

View Full Product Details