Product Details

SNP ID
rs200467282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42102277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTTTCTAGACAACTGCTTCACG[A/G]GGTAAAGCCACAATGCAGACAGAAA
Phenotype
MIM: 613859
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UMODL1 PubMed Links

Gene Details

Gene
UMODL1
Gene Name
uromodulin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004416.2 4222 Missense Mutation GAG,GGG E433G NP_001004416.2
NM_001199527.1 4222 Missense Mutation GAG,GGG E361G NP_001186456.1
NM_001199528.2 4222 Missense Mutation GAG,GGG E361G NP_001186457.2
NM_173568.3 4222 Missense Mutation GAG,GGG E433G NP_775839.3
XM_011529797.1 4222 Missense Mutation GAG,GGG E255G XP_011528099.1
XM_017028506.1 4222 Missense Mutation GAG,GGG E361G XP_016883995.1
XM_017028507.1 4222 Missense Mutation GAG,GGG E329G XP_016883996.1
XM_017028508.1 4222 Intron XP_016883997.1
Gene
UMODL1-AS1
Gene Name
UMODL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details