Product Details

SNP ID
rs201211615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30718743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCCACAGCTCCCACACACACTT[C/T]TGTAATTGAAATACATAGTGTCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP21-3 PubMed Links

Gene Details

Gene
KRTAP21-3
Gene Name
keratin associated protein 21-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164435.1 35 Missense Mutation AAA,AGA K6R NP_001157907.1

View Full Product Details