Product Details

SNP ID
rs200389479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30337200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGACACTGACATTGGCTAGATG[A/C]ACGTTCCAGAGTTTCGAAATTTTTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP27-1 PubMed Links

Gene Details

Gene
KRTAP27-1
Gene Name
keratin associated protein 27-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077711.1 495 Missense Mutation GCA,TCA A157S NP_001071179.1

View Full Product Details