Product Details

SNP ID
rs200661049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31105943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAAACTCACCTCCTTTAGGCGG[C/T]TCAGCTGTCATCCCCCGCAGGTCTG
Phenotype
MIM: 606481 MIM: 610918 MIM: 602127
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INPP5J PubMed Links

Gene Details

Gene
INPP5J
Gene Name
inositol polyphosphate-5-phosphatase J
There are no transcripts associated with this gene.

Gene
SELM
Gene Name
selenoprotein M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080430.2 212 Missense Mutation AAC,AGC N51S NP_536355.1
Gene
SMTN
Gene Name
smoothelin
There are no transcripts associated with this gene.

View Full Product Details