Product Details

SNP ID
rs201207849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42161350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTCCACCACCTTCTCATCTCCA[C/T]AGTCTCACCTGGAAGACAAGGGACA
Phenotype
MIM: 603107
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCF20 PubMed Links

Gene Details

Gene
TCF20
Gene Name
transcription factor 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005650.3 6257 UTR 3 NP_005641.1
NM_181492.2 6257 Silent Mutation CTA,CTG L1936L NP_852469.1
XM_005261722.3 6257 UTR 3 XP_005261779.1
XM_006724313.3 6257 UTR 3 XP_006724376.1
XM_011530353.2 6257 UTR 3 XP_011528655.1
XM_011530354.2 6257 Silent Mutation CTA,CTG L1936L XP_011528656.1

View Full Product Details